Learn More
Med Vet International Caring Cloth Silk Adhesive Tape, 1" X 10 Yds. Ea.
Supplier:  Med Vet International PRM260101HH
Encompass_Preferred
Sturdy rayon/acetate fabric helps secure dressings in place. Serrated edges facilitate tearing. Durable adhesion with minimal residue on skin. Water repellent and hypoallergenic.
Catalog No. 50-282-9406
 Restricted Item