Learn More
Med Vet International McKesson Cloth Medical Tape, 2 Inch x 10 Yard, White, 172-49220, 12/Case
Catalog No. 503044417
Encompass_Preferred
Medical Tape McKesson Perforated Soft Cloth 2 Inch X 10 Yard White NonSterile Breathable, spun-lace, nonwoven cloth can be used in a wide variety of surgical and medical applications Bidirectional tear with 2 inch perforations for easy tearing Medical-grade acrylic adhesive gently removes from the skin without leaving any residue behind Flexible design helps maintain the tape's shape to enhance secure placement and accommodate swelling
Catalog No. 50-304-4417 Supplier Med Vet International Supplier No. 1084092CS
Medical Tape McKesson Perforated Soft Cloth 2 Inch X 10 Yard White NonSterile Breathable, spun-lace, nonwoven cloth can be used in a wide variety of surgical and medical applications Bidirectional tear with 2 inch perforations for easy tearing Medical-grade acrylic adhesive gently removes from the skin without leaving any residue behind Flexible design helps maintain the tape's shape to enhance secure placement and accommodate swelling