Learn More
Med Vet International Coban Tape, 2" x 5 yds
Catalog No. 502522584
Encompass_Preferred
Self-adherent, non-adhesive elastic wrap, secures, protects, compresses, and supports. Maintains uniform compression. Won't slip or loosen. No need for clips or pins. Doesn't stick to hair or skin. Lightweight, porous, and comfortable. Moisture resistant. All rolls are 5 yards in length. No choice of colors. Coban® is a registered trademark of 3M. Can be used as vet wrap.
Catalog No. 50-252-2584 Supplier Med Vet International Supplier No. 1582
Self-adherent, non-adhesive elastic wrap, secures, protects, compresses, and supports. Maintains uniform compression. Won't slip or loosen. No need for clips or pins. Doesn't stick to hair or skin. Lightweight, porous, and comfortable. Moisture resistant. All rolls are 5 yards in length. No choice of colors. Coban® is a registered trademark of 3M. Can be used as vet wrap.