Learn More
Med Vet International Coban&Reg; Tape, 6" X 5 Yds.
Catalog No. 502829675
Encompass_Preferred
Self-adherent, non-adhesive elastic wrap, secures, protects, compresses, and supports. Maintains uniform compression. Won't slip or loosen. No need for clips or pins. Doesn't stick to hair or skin. Lightweight, porous, and comfortable. Moisture resistant. All rolls are 5 yards in length. No choice of colors. Coban; is a registered trademark of 3M. Can be used as vet wrap.
Catalog No. 50-282-9675 Supplier Med Vet International Supplier No. 1586
Self-adherent, non-adhesive elastic wrap, secures, protects, compresses, and supports. Maintains uniform compression. Won't slip or loosen. No need for clips or pins. Doesn't stick to hair or skin. Lightweight, porous, and comfortable. Moisture resistant. All rolls are 5 yards in length. No choice of colors. Coban; is a registered trademark of 3M. Can be used as vet wrap.