Learn More
Electron Microscopy Sciences XYZ-Axis Electrically Conductive Tape 9713, 4" wide 36 YD
SDP
Catalog No. 501901653
Encompass_Preferred
3M™ XYZ-Axis Electrically Conductive Tape 9713 is an isotropically conductive pressure sensitive tape that is ideal for scanning electron microscopy. 3M tape 9713 conducts electricity through the thickness (Z-axis) and in the plane of the adhesive (X, Y planes). The tape consists of a high performance 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance. The conductive fibers in 3M tape 9713 also provide improved handling characteristics.
Width: 4"
Catalog No. 50-190-1653 Supplier Electron Microscopy Sciences Supplier No. 77809100
3M™ XYZ-Axis Electrically Conductive Tape 9713 is an isotropically conductive pressure sensitive tape that is ideal for scanning electron microscopy. 3M tape 9713 conducts electricity through the thickness (Z-axis) and in the plane of the adhesive (X, Y planes). The tape consists of a high performance 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance. The conductive fibers in 3M tape 9713 also provide improved handling characteristics.
Width: 4"