Learn More
Electron Microscopy Sciences XYZ-Axis Electrically Conductive Tape 9712 | Double-Sided | 6.35mm x 32.9m
SDP
Catalog No. 501901651
Encompass_Preferred
Conductive Tape 9712 is an isotropically conductive pressure sensitive tape. The tape consists of a 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance with very high conductivity. The 9712 Tape is an excellent choice when needing to bond your samples for all SEM work.
Catalog No. 50-190-1651 Supplier Electron Microscopy Sciences Supplier No. 7780863
Conductive Tape 9712 is an isotropically conductive pressure sensitive tape. The tape consists of a 3M adhesive loaded with conductive fibers. The result is a double-sided tape providing both good adhesion and good electrical performance with very high conductivity. The 9712 Tape is an excellent choice when needing to bond your samples for all SEM work.