Learn More
Electron Microscopy Sciences Scotch™ Tape Dispenser
SDP
Catalog No. 5028518
Encompass_Preferred
Durable and economical; high impact resistant plastic dispenser. It features a permanently attached tape drum to eliminate loss. A rubber base prevents slipping. Two tone brown and beige. Holds tape up to 1" wide. 3" core.
Catalog No. 50-285-18 Supplier Electron Microscopy Sciences Supplier No. 77000
Durable and economical; high impact resistant plastic dispenser. It features a permanently attached tape drum to eliminate loss. A rubber base prevents slipping. Two tone brown and beige. Holds tape up to 1" wide. 3" core.