Learn More
Electron Microscopy Sciences Tape Dispenser Model MD-450, 20 Rolls, 3" Height
SDP
Catalog No. 501931305
Encompass_Preferred
For use with 60-yard tape rolls. Made from durable polycarbonate for visibility and easy cleaning. Cutting edge is stainless steel and suction feet help hold in place. Spindles drop in for quickly changing out rolls.
Rolls: 20
Height: 3"
Width: 13 ¼"
Depth: 6 ¼"
Catalog No. 50-193-1305 Supplier Electron Microscopy Sciences Supplier No. 7705305
For use with 60-yard tape rolls. Made from durable polycarbonate for visibility and easy cleaning. Cutting edge is stainless steel and suction feet help hold in place. Spindles drop in for quickly changing out rolls.
Rolls: 20
Height: 3"
Width: 13 ¼"
Depth: 6 ¼"