Learn More
Med Vet International Cover-Roll Stretch Nonwoven Polyester Dressing Retention Tape, 2 Inch x 10 Yard, White, 45552, 12/Case
Catalog No. 503043406
Encompass_Preferred
The soft, stretchable nonwoven polyester material adapts well to body contours Provides light compression without causing constriction or congestion The polyacrylate adhesive holds dressing safely and reliably in place, even in presence of moisture Radio-transparent to eliminate the need to remove for X-ray examinations 2 inches x 10 yards
Catalog No. 50-304-3406 Supplier Med Vet International Supplier No. 191703CS
The soft, stretchable nonwoven polyester material adapts well to body contours Provides light compression without causing constriction or congestion The polyacrylate adhesive holds dressing safely and reliably in place, even in presence of moisture Radio-transparent to eliminate the need to remove for X-ray examinations 2 inches x 10 yards