Learn More
Med Vet International Mefix Nonwoven Dressing Retention Tape, 4 Inch x 11 Yard, White, 311099, 1/Box
Catalog No. 503043715
Encompass_Preferred
This stretchable, nonabsorbent fabric dressing retention tape is adjustable enough to adhere to even flexible areas of the body The fabric prevents blood and other fluids from seeping to keep the area dry The 4-inch by 11-yard skin-friendly Mefix medical tape has a pre-measured and perforated backing The white, non-sterile, non-woven fabric is breathable but also provides containment of blood and bodily fluids The water-based, solvent-free adhesive in this Mefix dressing tape provides secure fixation with less irritation and more comfort
Catalog No. 50-304-3715 Supplier Med Vet International Supplier No. 684056BX
This stretchable, nonabsorbent fabric dressing retention tape is adjustable enough to adhere to even flexible areas of the body The fabric prevents blood and other fluids from seeping to keep the area dry The 4-inch by 11-yard skin-friendly Mefix medical tape has a pre-measured and perforated backing The white, non-sterile, non-woven fabric is breathable but also provides containment of blood and bodily fluids The water-based, solvent-free adhesive in this Mefix dressing tape provides secure fixation with less irritation and more comfort