Learn More
Med Vet International Omnifix Elastic Synthetic Rubber / Fiber Dressing Retention Tape, 2 Inch x 10 Yard, White, 900602, 63/Case
Catalog No. 503044929
Encompass_Preferred
Nonwoven retention tape Soft and conformable Highly permeable to air and moisture Helps reduce potential for maceration Provides reliable adherence Self-adhesive tape is 5 centimeters wide and 10 meters long
Catalog No. 50-304-4929 Supplier Med Vet International Supplier No. 1129500CS
Nonwoven retention tape Soft and conformable Highly permeable to air and moisture Helps reduce potential for maceration Provides reliable adherence Self-adhesive tape is 5 centimeters wide and 10 meters long