Learn More
Med Vet International Dressing Retention Tape with Liner Hypafix White 2 Inch X 10 Yard Nonwoven Polyester NonSterile, 4209, Each
Catalog No. 503043990
Encompass_Preferred
Hypafix dressing retention tape keeps dressings, absorbent pads and ostomy appliances in place without restricting movement Its non-woven, stretchable material offers flexibility and comfort This white dressing tape molds to different body contours for convenient access The wide-area fixation maximizes coverage to help prevent contamination A non-irritating adhesive makes the tape feel gentle on the skin The backing material includes a split liner to facilitate quick and easy application The non-sterile, skin-friendly tape resists water intrusion to maintain a secure fixation
Catalog No. 50-304-3990 Supplier Med Vet International Supplier No. 1065318RL
Hypafix dressing retention tape keeps dressings, absorbent pads and ostomy appliances in place without restricting movement Its non-woven, stretchable material offers flexibility and comfort This white dressing tape molds to different body contours for convenient access The wide-area fixation maximizes coverage to help prevent contamination A non-irritating adhesive makes the tape feel gentle on the skin The backing material includes a split liner to facilitate quick and easy application The non-sterile, skin-friendly tape resists water intrusion to maintain a secure fixation