Learn More
Med Vet International Hypafix Nonwoven Dressing Retention Tape, 2 Inch x 2 Yard, White, 4215, 36/Case
Catalog No. 503043968
Encompass_Preferred
This 2-inch x 2-yard Hypafix dressing retention tape keeps dressings, absorbent pads and ostomy appliances in place without restricting movement Its non-woven, stretchable material offers flexibility and comfort This white dressing retention sheet molds to different body contours for convenient access The wide-area fixation maximizes coverage to help prevent contamination A non-irritating adhesive makes the tape feel gentle on the skin The backing material includes a split liner to facilitate quick and easy application The non-sterile, skin-friendly tape resists water intrusion to maintain a secure fixation
Catalog No. 50-304-3968 Supplier Med Vet International Supplier No. 1065315CS
This 2-inch x 2-yard Hypafix dressing retention tape keeps dressings, absorbent pads and ostomy appliances in place without restricting movement Its non-woven, stretchable material offers flexibility and comfort This white dressing retention sheet molds to different body contours for convenient access The wide-area fixation maximizes coverage to help prevent contamination A non-irritating adhesive makes the tape feel gentle on the skin The backing material includes a split liner to facilitate quick and easy application The non-sterile, skin-friendly tape resists water intrusion to maintain a secure fixation