Learn More
Med Vet International 3M Durapore Medical Tape, White, 2 inch X 10 Yard, Silk-Like Cloth, Nonsterile, 1538-2, 6/Box
Catalog No. 502832175
Encompass_Preferred
3M Durapore white surgical tape is well-suited for securing high-strength applications The non-stretch silk-like cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin The material conforms to the body, letting you achieve a personalized fit Bi-directional tear for quick use Tape won't stick to gloves so tape can be torn while wearing gloves 2 inches x 10 yards
Catalog No. 50-283-2175 Supplier Med Vet International Supplier No. 5776BX
3M Durapore white surgical tape is well-suited for securing high-strength applications The non-stretch silk-like cloth retains its shape for reliable performance This hypoallergenic 3M Durapore medical tape is safe for sensitive skin The material conforms to the body, letting you achieve a personalized fit Bi-directional tear for quick use Tape won't stick to gloves so tape can be torn while wearing gloves 2 inches x 10 yards