Learn More
Med Vet International 3M Fiberglass Casting Tape, 2" X 4Yds, Each
Catalog No. 502830170
Encompass_Preferred
A lightweight, strong and durable casting tape that combines the benefits of fiberglass casting tape with the handling ease of plaster. Provides a durable cast with less chance of breakdown and need for costly repairs through secondary and tertiary casting. Made from water-activated resin which allows easier, tack-free application. High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and need for costly repairs or replacements. Lightweight for easy mobility Stretches to conform well to contours for a more comfortable Better-fitting cast Porous weave allows the skin to breathe No choice of color.
Catalog No. 50-283-0170 Supplier Med Vet International Supplier No. 3MFG2
A lightweight, strong and durable casting tape that combines the benefits of fiberglass casting tape with the handling ease of plaster. Provides a durable cast with less chance of breakdown and need for costly repairs through secondary and tertiary casting. Made from water-activated resin which allows easier, tack-free application. High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and need for costly repairs or replacements. Lightweight for easy mobility Stretches to conform well to contours for a more comfortable Better-fitting cast Porous weave allows the skin to breathe No choice of color.