Learn More
Med Vet International Fiberglass Casting Tape, 4" X 4Yds., Black, Each
Catalog No. 502831800
Encompass_Preferred
Strong, lightweight, high-quality fiberglass casting tape with water-activated polyurethane resin dries to a smooth finish so it's gentle against exposed skin. Easy-to-use formulation with fresh fragrance makes application a better experience for both staff and patients. Conforms well to contours for a more comfortable fit. Allows for penetration by X-rays to monitor healing.
Catalog No. 50-283-1800 Supplier Med Vet International Supplier No. FG4BLACKEA
Strong, lightweight, high-quality fiberglass casting tape with water-activated polyurethane resin dries to a smooth finish so it's gentle against exposed skin. Easy-to-use formulation with fresh fragrance makes application a better experience for both staff and patients. Conforms well to contours for a more comfortable fit. Allows for penetration by X-rays to monitor healing.