Learn More
Med Vet International Gas Plasma Indicator Tape 3M Comply 3/4 inch X 60 Yard Gas Plasma, 1228, 24/Case
Catalog No. 502830794
Encompass_Preferred
For use with STERRAD Sterilizers (Models 50, 100S & 200) A chemical indicator monitoring STERRAD Sterilizers (Models 50, 100S and 200) that verifies exposure to vapor hydrogen peroxide Gives the sterilizer operator assurance that each pack or item has been exposed to the sterilization process Consistent blue-to-pink color change for all 3M chemical indicators for vaporized hydrogen peroxide sterilization Ink does not contain lead Trusted, high quality adhesive
Catalog No. 50-283-0794 Supplier Med Vet International Supplier No. 456019CS
For use with STERRAD Sterilizers (Models 50, 100S & 200) A chemical indicator monitoring STERRAD Sterilizers (Models 50, 100S and 200) that verifies exposure to vapor hydrogen peroxide Gives the sterilizer operator assurance that each pack or item has been exposed to the sterilization process Consistent blue-to-pink color change for all 3M chemical indicators for vaporized hydrogen peroxide sterilization Ink does not contain lead Trusted, high quality adhesive