Learn More
Grainger Tape Gold 50 ft L 10 in W
Catalog No. 502749202
Encompass_Preferred
The Grainger Approved Gold Duct Tape is a 50 foot roll of 10 inch wide duct tape. It has a polyethylene coated cloth backing and a rubber adhesive. The tape is UL listed, conforms to UL 723, and is designed for HVAC and insulation applications. It can withstand temperatures up to 200°F. The tape has good adhesion, tensile strength, and elongation. It tears straight and resists abrasion. The tape can be used for sealing, patching, holding, protecting, and waterproofing.
Catalog No. 50-274-9202 Supplier Grainger Supplier No. 14A573
The Grainger Approved Gold Duct Tape is a 50 foot roll of 10 inch wide duct tape. It has a polyethylene coated cloth backing and a rubber adhesive. The tape is UL listed, conforms to UL 723, and is designed for HVAC and insulation applications. It can withstand temperatures up to 200°F. The tape has good adhesion, tensile strength, and elongation. It tears straight and resists abrasion. The tape can be used for sealing, patching, holding, protecting, and waterproofing.