Learn More
Grainger Tape Gray 50 ft L 10 in W
Catalog No. 502752741
Encompass_Preferred
The DID Gray Duct Tape is a 50 foot roll of 10 inch wide gray duct tape. It is made of polyethylene coated cloth with a rubber adhesive. The tape is designed for sealing, repairing, holding, and general purpose use. It can be used for HVAC, automotive, construction, industrial, and home applications. The tape has a temperature range of 50°F to 200°F. It conforms to UL 723 and ASTM D1000 standards. The tape has good adhesion, tensile strength, and tear resistance. It can be torn by hand without curling. The tape is lead and asbestos free.
Catalog No. 50-275-2741 Supplier Grainger Supplier No. 14A576
The DID Gray Duct Tape is a 50 foot roll of 10 inch wide gray duct tape. It is made of polyethylene coated cloth with a rubber adhesive. The tape is designed for sealing, repairing, holding, and general purpose use. It can be used for HVAC, automotive, construction, industrial, and home applications. The tape has a temperature range of 50°F to 200°F. It conforms to UL 723 and ASTM D1000 standards. The tape has good adhesion, tensile strength, and tear resistance. It can be torn by hand without curling. The tape is lead and asbestos free.