Learn More
Med Vet International Waterproof Medical Tape Hy-Tape Pink 1 Inch X 5 Yard Zinc Oxide Adhesive Zinc Oxide NonSterile, 10LF, 12/Tube
Catalog No. 503044755
Encompass_Preferred
Water-resistant, washable, plastic tapes have a zinc oxide adhesive base They are flexible and easy to remove and can adhere even to wet or oily skin Hy-Tape tapes' protective nature is ideal for extended wear or when it will come into contact with moisture of any type Pink zinc oxide adhesive medical tape is 1 inch wide and 5 yards long
Catalog No. 50-304-4755 Supplier Med Vet International Supplier No. 1190129TU
Water-resistant, washable, plastic tapes have a zinc oxide adhesive base They are flexible and easy to remove and can adhere even to wet or oily skin Hy-Tape tapes' protective nature is ideal for extended wear or when it will come into contact with moisture of any type Pink zinc oxide adhesive medical tape is 1 inch wide and 5 yards long