Learn More
Med Vet International Waterproof Medical Tape Hy-Tape Pink 1/2 Inch X 5 Yard Zinc Oxide Adhesive Zinc Oxide NonSterile, 105BLF, Each
Catalog No. 503043158
Encompass_Preferred
Water-resistant, washable, plastic tapes have a zinc oxide adhesive base They are flexible and easy to remove and can adhere even to wet or oily skin Hy-Tape tapes' protective nature is ideal for extended wear or when it will come into contact with moisture of any type
Catalog No. 50-304-3158 Supplier Med Vet International Supplier No. 467563EA
Water-resistant, washable, plastic tapes have a zinc oxide adhesive base They are flexible and easy to remove and can adhere even to wet or oily skin Hy-Tape tapes' protective nature is ideal for extended wear or when it will come into contact with moisture of any type