Learn More
Med Vet International Steam Indicator Tape 3M Comply 1 inch X 60 Yard Steam, 1355-24mm, 20/Case
Catalog No. 502831523
Encompass_Preferred
For use as a process indicator to show that a pack has been exposed to the steam sterilization process Also serves to seal sterilization packs Can be used on disposable and reusable wraps Lead free ink means no hazardous waste issues Trusted, high quality adhesive Stretchable backing that minimizes tape "pop-off" during sterilization Can be written on or labeled with pre-printed labels
Catalog No. 50-283-1523 Supplier Med Vet International Supplier No. 775750CS
For use as a process indicator to show that a pack has been exposed to the steam sterilization process Also serves to seal sterilization packs Can be used on disposable and reusable wraps Lead free ink means no hazardous waste issues Trusted, high quality adhesive Stretchable backing that minimizes tape "pop-off" during sterilization Can be written on or labeled with pre-printed labels