Learn More
Med Vet International Steam Indicator Tape 3M Comply 1 inch X 60 Yard Steam, 1322-24mm, 20/Case
Catalog No. 502829562
Encompass_Preferred
3M's Comply lead-free steam indicator tape has a chemical indicator ink that helps monitor steam sterilization Reliable adhesive seals sterilization packs The ink is lead-free to avoid hazardous waste issues The sterilization indicator tape stretches to avoid disruption during sterilization 1 inch x 60 yards
Catalog No. 50-282-9562 Supplier Med Vet International Supplier No. 775747CS
3M's Comply lead-free steam indicator tape has a chemical indicator ink that helps monitor steam sterilization Reliable adhesive seals sterilization packs The ink is lead-free to avoid hazardous waste issues The sterilization indicator tape stretches to avoid disruption during sterilization 1 inch x 60 yards