Learn More
Med Vet International Steam Indicator Tape 3M Comply 3/4 inch X 60 Yard Steam, 1322-18mm, 28/Case
Catalog No. 502831506
Encompass_Preferred
For use as a process indicator to show that a pack has been exposed to the steam sterilization process Also serves to seal sterilization packs Can be used on disposable and reusable wraps Lead free ink means no hazardous waste issues Trusted, high quality adhesive Stretchable backing that minimizes tape "pop-off" during sterilization Can be written on or labeled with pre-printed labels 3/4 inch x 60 yards
Catalog No. 50-283-1506 Supplier Med Vet International Supplier No. 775746CS
For use as a process indicator to show that a pack has been exposed to the steam sterilization process Also serves to seal sterilization packs Can be used on disposable and reusable wraps Lead free ink means no hazardous waste issues Trusted, high quality adhesive Stretchable backing that minimizes tape "pop-off" during sterilization Can be written on or labeled with pre-printed labels 3/4 inch x 60 yards