Learn More
Med Vet International McKesson Steam Indicator Tape, 1 Inch x 60 Yard, 73-BT36, 18/Case
Catalog No. 503044636
Encompass_Preferred
Steam Indicator Tape McKesson 1 Inch X 60 Yard Steam Blue Blue steam tape has a more aggressive adhesive and is for disposable wraps only For use in securing wrapped instrument sets before, during and after sterilization Each roll of McKesson tape is individually packaged to ensure quality at time of use 1 inch x 60 yards
Catalog No. 50-304-4636 Supplier Med Vet International Supplier No. 524891CS
Steam Indicator Tape McKesson 1 Inch X 60 Yard Steam Blue Blue steam tape has a more aggressive adhesive and is for disposable wraps only For use in securing wrapped instrument sets before, during and after sterilization Each roll of McKesson tape is individually packaged to ensure quality at time of use 1 inch x 60 yards