Learn More
MSC Tape Logic Anti-Slip Tape, Black, 1 in Wide, 1 Roll
Catalog No. 5000803844
Encompass
This anti-slip tape helps prevent slipping and falling. It features an 80 grit abrasive sandpaper surface with an adhesive backing, suitable for non-porous surfaces like sealed concrete, painted metals, and wood.
  • Helps prevent slipping and falling
  • Abrasive sandpaper surface, 80 grit
  • Moisture and grease resistant
  • Suitable for temperatures from -20° to 150° F
  • Heavy-duty with coarser, thicker surface
  • Aggressive adhesive for enhanced protection
  • Reflective properties
Catalog No. 50-008-03844 Supplier MSC Supplier No. 16006058
This anti-slip tape helps prevent slipping and falling. It features an 80 grit abrasive sandpaper surface with an adhesive backing, suitable for non-porous surfaces like sealed concrete, painted metals, and wood.
  • Helps prevent slipping and falling
  • Abrasive sandpaper surface, 80 grit
  • Moisture and grease resistant
  • Suitable for temperatures from -20° to 150° F
  • Heavy-duty with coarser, thicker surface
  • Aggressive adhesive for enhanced protection
  • Reflective properties