Learn More
MSC Tape Logic Labels, Paper, 4-1/4 x 7 in, 1 ea
Catalog No. 5000788106
Encompass
These white paper laser labels are designed for creating custom product or mailing labels. They are compatible with both laser and inkjet printers and adhere to a variety of smooth surfaces.
  • Overall dimensions of 4-1/4 in length and 7 in width
  • Suitable for laser and inkjet printers
  • Compatible with paper, corrugated materials, wood, glass, plastic, and metal surfaces
Catalog No. 50-007-88106 Supplier MSC Supplier No. 10242543
These white paper laser labels are designed for creating custom product or mailing labels. They are compatible with both laser and inkjet printers and adhere to a variety of smooth surfaces.
  • Overall dimensions of 4-1/4 in length and 7 in width
  • Suitable for laser and inkjet printers
  • Compatible with paper, corrugated materials, wood, glass, plastic, and metal surfaces