Learn More
Med Vet International McKesson Cloth Tape Measure, 72", 63-4414, 6/Box
Supplier:  Med Vet International 481650BX
Encompass_Preferred
Measurement Tape McKesson 72 Inch Cloth Reusable English / Metric White plastic case with push button to retract tape measure Reusable Durable linen-like fiberglass tape measure 72L X 1/4W Inch Retractable Cloth Graduated English Scale & Metric Scale on reverse side
Catalog No. 50-304-4087