Learn More
Med Vet International Perforated Medical Tape 3M Medipore H White 3 inch X 10 Yard Soft Cloth Nonsterile, 2863, 12/Case
Catalog No. 502829476
Encompass_Preferred
The water-resistant, non-sterile 3M Medipore H tape is suitable for securing dressings, tubing and catheters The multi-directional stretch white fabric allows for freedom of movement and wound swelling This 3-inch x 10-yard 3M Medipore H soft cloth tape includes perforations for quick tearing by hand The porous, breathable cloth material allows for aeration while remaining gentle on the skin It is latex-free and hypoallergenic for those with sensitive skin
Catalog No. 50-282-9476 Supplier Med Vet International Supplier No. 324082CS
The water-resistant, non-sterile 3M Medipore H tape is suitable for securing dressings, tubing and catheters The multi-directional stretch white fabric allows for freedom of movement and wound swelling This 3-inch x 10-yard 3M Medipore H soft cloth tape includes perforations for quick tearing by hand The porous, breathable cloth material allows for aeration while remaining gentle on the skin It is latex-free and hypoallergenic for those with sensitive skin