Learn More
Med Vet International Perforated Medical Tape 3M Medipore H White 4 inch X 10 Yard Soft Cloth Nonsterile, 2864, 12/Case
Catalog No. 502831382
Encompass_Preferred
This 4-inch x 10-yard 3M Medipore H medical tape is suitable for securing dressings, tubing and catheters The multi-directional stretch fabric allows for freedom of movement and wound swelling 3M Medipore H soft tape cloth includes perforations for quick tearing by hand The porous, breathable material allows for aeration while remaining gentle on the skin
Catalog No. 50-283-1382 Supplier Med Vet International Supplier No. 314412CS
This 4-inch x 10-yard 3M Medipore H medical tape is suitable for securing dressings, tubing and catheters The multi-directional stretch fabric allows for freedom of movement and wound swelling 3M Medipore H soft tape cloth includes perforations for quick tearing by hand The porous, breathable material allows for aeration while remaining gentle on the skin