Learn More
Med Vet International Perforated Medical Tape 3M Medipore H White 8 inch X 10 Yard Soft Cloth Nonsterile, 2868, 6/Case
Catalog No. 502830237
Encompass_Preferred
This 8-inch x 10-yard 3M Medipore H medical tape is suitable for securing dressings, tubing and catheters The multi-directional stretch fabric allows for freedom of movement and wound swelling 3M Medipore H soft tape cloth includes perforations for quick tearing by hand The porous, breathable material allows for aeration while remaining gentle on the skin There are six rolls of white medical tape in a case
Catalog No. 50-283-0237 Supplier Med Vet International Supplier No. 342586CS
This 8-inch x 10-yard 3M Medipore H medical tape is suitable for securing dressings, tubing and catheters The multi-directional stretch fabric allows for freedom of movement and wound swelling 3M Medipore H soft tape cloth includes perforations for quick tearing by hand The porous, breathable material allows for aeration while remaining gentle on the skin There are six rolls of white medical tape in a case