Learn More
Med Vet International 3M Micropore Medical Tape, White, 1/2 inch X 10 Yard, Paper, Nonsterile, 1530-0, 240/Case
Catalog No. 502832370
Encompass_Preferred
3M Micropore medical tape has a quality adhesive for good adhesion to skin It leaves minimal adhesive residue for quick removal The white tape is water-resistant and can adhere to damp surfaces It conforms well to cover hard to dress areas The paper tape is hypoallergenic and safe to use on sensitive skin This tape is known in hospitals as 3M Micropore surgical tape It's highly breathable to help maintain proper skin condition White tape is 1/2 inch x 10 yards
Catalog No. 50-283-2370 Supplier Med Vet International Supplier No. 5766CS
3M Micropore medical tape has a quality adhesive for good adhesion to skin It leaves minimal adhesive residue for quick removal The white tape is water-resistant and can adhere to damp surfaces It conforms well to cover hard to dress areas The paper tape is hypoallergenic and safe to use on sensitive skin This tape is known in hospitals as 3M Micropore surgical tape It's highly breathable to help maintain proper skin condition White tape is 1/2 inch x 10 yards