Learn More
Med Vet International 3M Micropore Paper Medical Tape, 1in X 10 Yard, White, 1530-1, 120/Case
Catalog No. 502830868
Encompass_Preferred
Medical Tape 3M™ Micropore™ White 1 Inch X 10 Yard Paper NonSterile 3M Micropore medical tape has a quality adhesive for good adhesion to the skin Leaves minimal adhesive residue for quick removal White tape is water-resistant and can adhere to damp surfaces Conforms well to cover hard to dress areas Hypoallergenic and safe to use on sensitive skin Highly breathable to help maintain proper skin condition This tape is known in hospitals as 3M Micropore surgical tape Each box has 12 rolls of 1 inch x 10 yard medical tape
Catalog No. 50-283-0868 Supplier Med Vet International Supplier No. 5767CS
Medical Tape 3M™ Micropore™ White 1 Inch X 10 Yard Paper NonSterile 3M Micropore medical tape has a quality adhesive for good adhesion to the skin Leaves minimal adhesive residue for quick removal White tape is water-resistant and can adhere to damp surfaces Conforms well to cover hard to dress areas Hypoallergenic and safe to use on sensitive skin Highly breathable to help maintain proper skin condition This tape is known in hospitals as 3M Micropore surgical tape Each box has 12 rolls of 1 inch x 10 yard medical tape