Learn More
Med Vet International 3M Micropore Medical Tape, Small, Blue, 1 inch X 5-1/2 Yard, Silicone, Nonsterile, 2770-1, 120/Case
Catalog No. 502831413
Encompass_Preferred
3M™ Micropore™ S surgical tape comes off cleanly without causing pain or disturbing the wound healing process A strong adhesive forms a secure bond to the skin for reliable protection Roll tears, with no scissors required, for ease of use Medical tape offers gentle wear, making it beneficial for those with skin sensitivities or compromised skin 2.5 centimeters x 5 meters
Catalog No. 50-283-1413 Supplier Med Vet International Supplier No. 774188CS
3M™ Micropore™ S surgical tape comes off cleanly without causing pain or disturbing the wound healing process A strong adhesive forms a secure bond to the skin for reliable protection Roll tears, with no scissors required, for ease of use Medical tape offers gentle wear, making it beneficial for those with skin sensitivities or compromised skin 2.5 centimeters x 5 meters