Learn More
Med Vet International 3M Micropore Paper Medical Tape, 2in X 10 Yard, White, 1530-2, 60/Case
Catalog No. 502829287
Encompass_Preferred
Medical Tape 3M™ Micropore™ White 2 Inch X 10 Yard Paper NonSterile 3M Micropore medical tape has a quality adhesive for good adhesion to skin It leaves minimal adhesive residue for quick removal The white tape is water-resistant and can adhere to damp surfaces It conforms well to cover hard to dress areas The Micropore paper tape is hypoallergenic and safe to use on sensitive skin It's highly breathable to help maintain proper skin condition This tape is known in hospitals as 3M Micropore surgical tape Each box has six rolls of 2 inch x 10 yard medical tape
Catalog No. 50-282-9287 Supplier Med Vet International Supplier No. 684272CS
Medical Tape 3M™ Micropore™ White 2 Inch X 10 Yard Paper NonSterile 3M Micropore medical tape has a quality adhesive for good adhesion to skin It leaves minimal adhesive residue for quick removal The white tape is water-resistant and can adhere to damp surfaces It conforms well to cover hard to dress areas The Micropore paper tape is hypoallergenic and safe to use on sensitive skin It's highly breathable to help maintain proper skin condition This tape is known in hospitals as 3M Micropore surgical tape Each box has six rolls of 2 inch x 10 yard medical tape