Learn More
Med Vet International Medical Tape Nexcare Gentle White 1 inch X 10 Yard Paper Nonsterile, 781-2Pk, 48/Box
Catalog No. 502830306
Encompass_Preferred
Nexcare gentle paper tape has a gentle adhesive that won't irritate the skin It is water-resistant to ensure it adheres when in contact with water or other liquids It conforms easily, allowing it to flex with movement The medical tape 1-inch x 10-yard roll is breathable to condition the skin It tears by hand for a quick and hassle-free application Nexcare first aid tape is hypoallergenic, ensuring it remains gentle on sensitive skin Two tape rolls in each pack
Catalog No. 50-283-0306 Supplier Med Vet International Supplier No. 1084004BX
Nexcare gentle paper tape has a gentle adhesive that won't irritate the skin It is water-resistant to ensure it adheres when in contact with water or other liquids It conforms easily, allowing it to flex with movement The medical tape 1-inch x 10-yard roll is breathable to condition the skin It tears by hand for a quick and hassle-free application Nexcare first aid tape is hypoallergenic, ensuring it remains gentle on sensitive skin Two tape rolls in each pack