Learn More
Med Vet International Perivaginal Tape, 5m., J0025
Catalog No. 5000495572
Encompass_Preferred
To be used according to the Dr. Buhner method for treatment of a prolapse of the uterus or vagina. This special woven oval-shaped tape has histophilic features that permit a heal-in without irritation. Directions for use upon request. 4.5 meters.
Catalog No. 50-004-95572 Supplier Med Vet International Supplier No. JOR25
To be used according to the Dr. Buhner method for treatment of a prolapse of the uterus or vagina. This special woven oval-shaped tape has histophilic features that permit a heal-in without irritation. Directions for use upon request. 4.5 meters.