Learn More
Grainger Tape Phosphorescent 33 ft L 10 in W
Catalog No. 502749225
Encompass_Preferred
The Grainger Approved Phosphorescent Tape is a self-adhesive glow-in-the-dark tape that provides low-level illumination in dark environments. It absorbs light during the day or when illuminated, storing the energy to glow green in the dark for up to 10 hours. This polyester film tape has an aggressive acrylic adhesive and is 0.5 in. wide x 30 ft. long. It has a temperature range of 32°F to 150°F. The phosphorescent tape creates maximum nighttime visibility on stairs, walkways, machinery, signage, and more. It meets military specification MIL-T-47113 and is made in the USA.
Catalog No. 50-274-9225 Supplier Grainger Supplier No. 14A561
The Grainger Approved Phosphorescent Tape is a self-adhesive glow-in-the-dark tape that provides low-level illumination in dark environments. It absorbs light during the day or when illuminated, storing the energy to glow green in the dark for up to 10 hours. This polyester film tape has an aggressive acrylic adhesive and is 0.5 in. wide x 30 ft. long. It has a temperature range of 32°F to 150°F. The phosphorescent tape creates maximum nighttime visibility on stairs, walkways, machinery, signage, and more. It meets military specification MIL-T-47113 and is made in the USA.