Learn More
Grainger Tape Purple 50 ft L 10 in W
Catalog No. 502749203
Encompass_Preferred
The Grainger 14A574 is a 14 gauge, 574 foot spool of electrical wire. It is UL listed type THHN wire with a 600 volt insulation rating. The wire has a stranded copper conductor and a PVC insulation and jacket. It can be used for general purpose wiring in conduit, cable trays or other approved raceways. This wire is suitable for both indoor and outdoor applications. The spool has an over-all diameter of 18 inches and a 5 inch arbor hole to mount on wire dispensing equipment.
Catalog No. 50-274-9203 Supplier Grainger Supplier No. 14A574
The Grainger 14A574 is a 14 gauge, 574 foot spool of electrical wire. It is UL listed type THHN wire with a 600 volt insulation rating. The wire has a stranded copper conductor and a PVC insulation and jacket. It can be used for general purpose wiring in conduit, cable trays or other approved raceways. This wire is suitable for both indoor and outdoor applications. The spool has an over-all diameter of 18 inches and a 5 inch arbor hole to mount on wire dispensing equipment.