Learn More
Cole-Parmer Extech 461937 Tachometer Reflective Tape 23"roll - 10 rolls/pk
Catalog No. 50993106 Shop All Cole-Parmer Products
Encompass
Tachometer optical reflective tape provides good reflective signal for any optical (laser or standard) tachometer. Package contains 10 - 23" long strips of highly reflective tape.
Catalog No. 50-993-106 Supplier Cole-Parmer Supplier No. 8770262
Tachometer optical reflective tape provides good reflective signal for any optical (laser or standard) tachometer. Package contains 10 - 23" long strips of highly reflective tape.