Learn More
Med Vet International McKesson Nonwoven Fabric / Printed Release Paper Dressing Retention Tape, 6 Inch x 10 Yard, White, 16-4806, 1/Box
Catalog No. 503044722
Encompass_Preferred
Dressing Retention Tape with Liner McKesson Water Resistant Nonwoven / Printed Release Paper 6 Inch X 10 Yard White NonSterile Acrylic adhesive strips on both sides Conforms to body contours Breathable, wear-resistant fabric Non-irritating adhesive Grid guidelines for easier measurement Printed released paper backing Packaged in a convenient dispenser box for quick, easy access
Catalog No. 50-304-4722 Supplier Med Vet International Supplier No. 1087973BX
Dressing Retention Tape with Liner McKesson Water Resistant Nonwoven / Printed Release Paper 6 Inch X 10 Yard White NonSterile Acrylic adhesive strips on both sides Conforms to body contours Breathable, wear-resistant fabric Non-irritating adhesive Grid guidelines for easier measurement Printed released paper backing Packaged in a convenient dispenser box for quick, easy access