Learn More
Med Vet International Cast Tape 3M Scotchcast Soft Cast 2 inch X 12 Foot Fiberglass Blue, 82102B, 10/Case
Catalog No. 502828944
Encompass_Preferred
Flexible, Soft Cast allows for mobility to accommodate specific patient requirements Easy to apply and remove for fast application and enhanced patient comfort Soft, pliable cast edges improve patient comfort Ideal for use in combination with rigid Scotchcast casting tape, as doft edges provide additional comfort 10 Rolls per Box
Catalog No. 50-282-8944 Supplier Med Vet International Supplier No. 447728CS
Flexible, Soft Cast allows for mobility to accommodate specific patient requirements Easy to apply and remove for fast application and enhanced patient comfort Soft, pliable cast edges improve patient comfort Ideal for use in combination with rigid Scotchcast casting tape, as doft edges provide additional comfort 10 Rolls per Box