Learn More
Med Vet International Cast Tape 3M Scotchcast Plus 2 inch X 12 Foot Fiberglass / Resin Bright Orange, 82002W, 10/Case
Catalog No. 502832137
Encompass_Preferred
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe 2 inches x 12 feet
Catalog No. 50-283-2137 Supplier Med Vet International Supplier No. 199238CS
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe 2 inches x 12 feet