Learn More
Med Vet International Cast Tape 3M Scotchcast Soft Cast 3 inch X 12 Foot Fiberglass Blue, 82103B, 1 Roll
Catalog No. 502830206
Encompass_Preferred
Flexible, Soft Cast allows for mobility to accommodate specific patient requirements Easy to apply and remove for fast application and enhanced patient comfort Soft, pliable cast edges improve patient comfort Ideal for use in combination with rigid Scotchcast casting tape, as doft edges provide additional comfort 10 Rolls per Box
Catalog No. 50-283-0206 Supplier Med Vet International Supplier No. 443501RL
Flexible, Soft Cast allows for mobility to accommodate specific patient requirements Easy to apply and remove for fast application and enhanced patient comfort Soft, pliable cast edges improve patient comfort Ideal for use in combination with rigid Scotchcast casting tape, as doft edges provide additional comfort 10 Rolls per Box