Learn More
Med Vet International Cast Tape 3M Scotchcast Soft Cast 3 inch X 12 Foot Fiberglass White, 82103, 10/Case
Catalog No. 502830783
Encompass_Preferred
Flexible, soft cast allows for mobility The soft, pliable cast edges improve comfort Can be used in combination with rigid fiberglass splints, as soft edges provide additional comfort White 3 inches x 12 feet
Catalog No. 50-283-0783 Supplier Med Vet International Supplier No. 198741CS
Flexible, soft cast allows for mobility The soft, pliable cast edges improve comfort Can be used in combination with rigid fiberglass splints, as soft edges provide additional comfort White 3 inches x 12 feet