Learn More
Med Vet International Cast Tape 3M Scotchcast Plus 1 X 72 inch Fiberglass / Resin White, 82001, 1 Roll
Catalog No. 502830634
Encompass_Preferred
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe White 1 inch x 6 feet
Catalog No. 50-283-0634 Supplier Med Vet International Supplier No. 979847RL
Water-activated resin lets you control working time according to application needs High and early strength allows for weight bearing in 20 minutes and provides a durable cast with less chance of breakdown and the need for costly repairs or replacements Lightweight for easy mobility Stretches to conform well to contours for a more comfortable, better fitting cast Porous weave allows the skin to breathe White 1 inch x 6 feet