Learn More
Med Vet International McKesson Silicone Medical Tape, 1 Inch x 5-1/2 Yard, Transparent, 16-48410, 12/Box
Catalog No. 503044434
Encompass_Preferred
Medical Tape McKesson Porous Plastic / Silicone 1 Inch X 5-1/2 Yard Transparent NonSterile Ideal for individuals with sensitive, compromised or at-risk skin who are concerned about minimizing pain Porous, conformable design offers secure placement and gentle wear Silicone adhesive can be removed cleanly without disrupting fragile skin layers Can be repositioned when repeated taping is necessary
Catalog No. 50-304-4434 Supplier Med Vet International Supplier No. 1073806BX
Medical Tape McKesson Porous Plastic / Silicone 1 Inch X 5-1/2 Yard Transparent NonSterile Ideal for individuals with sensitive, compromised or at-risk skin who are concerned about minimizing pain Porous, conformable design offers secure placement and gentle wear Silicone adhesive can be removed cleanly without disrupting fragile skin layers Can be repositioned when repeated taping is necessary