Learn More
Med Vet International Medical Tape 3M Micropore S Blue 1 inch X 1-1/2 Yard Silicone Nonsterile, 2770S-1, 100/Box
Catalog No. 502831775
Encompass_Preferred
Blue medical tape is 1 inch x 1-1/2 yards and works well at any length you need: Simply tear each strip with fingertips Offers short-term and long-term (48 hour) reliable and pliable fixation Comfort: Removes cleanly, without disrupting fragile skin layers or causing patients undue pain Ease of use: Can be repositioned and neatly torn by hand Helps reduce risk: Individually packaged single-use rolls can help protect patients by reducing the risk of cross-contamination
Catalog No. 50-283-1775 Supplier Med Vet International Supplier No. 774835BX
Blue medical tape is 1 inch x 1-1/2 yards and works well at any length you need: Simply tear each strip with fingertips Offers short-term and long-term (48 hour) reliable and pliable fixation Comfort: Removes cleanly, without disrupting fragile skin layers or causing patients undue pain Ease of use: Can be repositioned and neatly torn by hand Helps reduce risk: Individually packaged single-use rolls can help protect patients by reducing the risk of cross-contamination