Learn More
Med Vet International Steam Autoclave Tape, 3/4" X 60Yds.
Catalog No. 502829706
Encompass_Preferred
Steam autoclave tape is placed on outside of bag or instrument wrap and Indicates when sterilization is complete. Autoclave tape is an adhesive tape used in autoclaving (heating under high pressure steam to sterilize) to indicate whether a specific temperature has been reached. Autoclave tape works by changing color after exposure to temperatures in the sterilization processes, typically 249.°F
Catalog No. 50-282-9706 Supplier Med Vet International Supplier No. AUTOTAPE3/4
Steam autoclave tape is placed on outside of bag or instrument wrap and Indicates when sterilization is complete. Autoclave tape is an adhesive tape used in autoclaving (heating under high pressure steam to sterilize) to indicate whether a specific temperature has been reached. Autoclave tape works by changing color after exposure to temperatures in the sterilization processes, typically 249.°F