Learn More
Med Vet International Water Resistant Dressing Retention Tape With Liner 3M Multipore Dry White 1/2 inch X 5-1/2 Yard Pique Nonsterile, 3730-0, 1 Roll
Catalog No. 502831400
Encompass_Preferred
3M medical tape with bi-directional stretch conforms to your body's natural shape to give you reliable protection The water-resistant material is suitable for use on wet skin and damp body parts It's hypoallergenic for improved safety The grid pattern on the liner allows for quick and precise cuts in any shape or size 3M Multipore dry surgical tape is useful when dressing wounds or securing medical equipment like catheters or tubing
Catalog No. 50-283-1400 Supplier Med Vet International Supplier No. 1019017RL
3M medical tape with bi-directional stretch conforms to your body's natural shape to give you reliable protection The water-resistant material is suitable for use on wet skin and damp body parts It's hypoallergenic for improved safety The grid pattern on the liner allows for quick and precise cuts in any shape or size 3M Multipore dry surgical tape is useful when dressing wounds or securing medical equipment like catheters or tubing